Order Entry
Switzerland
ContactUsLinkComponent
 
VWR®, Wipes for General Applications
  115-0202
undefined
VWR®, Wipes for General Applications
  115-0202

 

  • Packed:
    In box
  • Description:
    Tissue wipes, 1 ply
  • L×W:
    213×114 mm
  • Pk:
    280

 

 

Absorbent, soft, thick, low lint tissues made from virgin fibres. For light use in the laboratory, electronics, computer industry, pharmaceuticals, biotechnology and optics.

  • High absorbency, in three formats, available in single or triple ply
  • Practical dispenser box with anti-static, plastic film window for removal
  • Table and wall-mounted dispensers for one handed removal of the wipes are available separately

Colour: White

Dispensers