Orders LinkContactUsLinkComponent
Vials with screw thread, Nalgene®
   Sustainable Options Available
Vials with screw thread, Nalgene®
   Sustainable Options Available
Certificates
Vials with screw thread, Nalgene®
7 Options Available Below
Product Details & Documents
These vials are ideal for diagnostic and biological sera, pharmaceuticals for veterinary and ophthalmologic usage as well as many other applications. They are made from clear or coloured brown PETG.
  • Available clear or coloured brown, sterile or non-sterile
  • A wide range of different screw caps is optionally available
  • Manufactured and packed in workrooms equipped with HEPA filters
Closures for vials: HDPE, coloured. For safe and tight closure of narrow-neck vials. Available in non-sterile or sterile versions. Non sterile caps are loose packed, sterile caps are loose packed in a box with a three-section insert.
For safe and tight closure of narrow-neck vials.
Specifications for Product Family
Specifications
  • Catalog No:
  • 215-1499
  • 215-1486
  • 215-1487
  • 215-1488
  • 215-1497
  • 215-1498
  • 215-1505
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
  • NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
  • NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
  • NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Vials without closures
Product Information
  • Capacity
    Colour
    Cap included
    Cap Size
    Sterile
    Availability
    Price
  • 215-1499
    Serum vial
    Serum vial
    20,00 ml
    Clear
    No
    20-415
    +
    Log in for availability
    Log in for availability
    1 * 612 ST
    1 * 612 ST - SFr. 579.00
    SFr. 579.00
    1 * 612 ST
       SFr. 579.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1499
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 215-1486
    Serum vial
    Serum vial
    5,00 ml
    Clear
    No
    20-415
    -
    Log in for availability
    Log in for availability
    1 * 1.656 ST
    1 * 1.656 ST - SFr. 1’100.00
    SFr. 1’100.00
    1 * 1.656 ST
       SFr. 1’100.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1486
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 215-1487
    Serum vial
    Serum vial
    10,00 ml
    Clear
    No
    20-415
    -
    Log in for availability
    Log in for availability
    1 * 1.260 ST
    1 * 1.260 ST - SFr. 898.00
    SFr. 898.00
    1 * 1.260 ST
       SFr. 898.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1487
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1488
    Serum vial
    Serum vial
    20,00 ml
    Clear
    No
    20-415
    -
    Log in for availability
    Log in for availability
    1 * 612 ST
    1 * 612 ST - SFr. 555.00
    SFr. 555.00
    1 * 612 ST
       SFr. 555.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1488
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 215-1497
    Serum vial
    Serum vial
    5,00 ml
    Clear
    No
    20-415
    +
    Log in for availability
    Log in for availability
    1 * 1.656 ST
    1 * 1.656 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1.656 ST
       SFr. 1’130.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1497
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • Serum vial
    10,00 ml
    Clear
    No
    20-415
    +
    Log in for availability
    Log in for availability
    1 * 1.260 ST
    1 * 1.260 ST - SFr. 939.00
    SFr. 939.00
    1 * 1.260 ST
       SFr. 939.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1498
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 215-1505
    Serum vial
    Serum vial
    10,00 ml
    Amber
    No
    20-415
    +
    Log in for availability
    Log in for availability
    1 * 1.260 ST
    1 * 1.260 ST - SFr. 1’040.00
    SFr. 1’040.00
    1 * 1.260 ST
       SFr. 1’040.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1505
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
Closures for vials
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Closures lined, white, sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 2’270.00
    SFr. 2’270.00
    1 * 2.000 ST
       SFr. 2’270.00
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Closures lined, white, non sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 2’190.00
    SFr. 2’190.00
    1 * 2.000 ST
       SFr. 2’190.00
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Closures lined, black, non sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 2’220.00
    SFr. 2’220.00
    1 * 2.000 ST
       SFr. 2’220.00
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Closures lined, blue, sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 2’410.00
    SFr. 2’410.00
    1 * 2.000 ST
       SFr. 2’410.00
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Closures lined, red, sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 2’410.00
    SFr. 2’410.00
    1 * 2.000 ST
       SFr. 2’410.00
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Closures lined, green, sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 2’300.00
    SFr. 2’300.00
    1 * 2.000 ST
       SFr. 2’300.00
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Closures lined, yellow, sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 2’300.00
    SFr. 2’300.00
    1 * 2.000 ST
       SFr. 2’300.00
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Closures lined, black, sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 2’300.00
    SFr. 2’300.00
    1 * 2.000 ST
       SFr. 2’300.00