Specifiche
- Description:Micro-insert 0,35 ml for Deepwell without cap
- Seq:09
- Environmentally Preferable:
- Pk:960
Specifiche
Informazioni su questo articolo
96-position systems (Ritter) with glass inserts are used in HTS (high throughput screening) chemistry when the pure PP block is not inert enough. The inserts can be sealed with a PE cap seal, or over the whole block with a Sealmat block cover made of silicone, laminated with a PTFE film.