CryoFlex™ sleeves for CryoLine™ tubes, Nunc™
Catalog # 479-6858
Supplier: Thermo Fisher Scientific
Specifications
- For:CryoLine™ tubes, Nunc™
- Description:CryoFlex™ sleeves
- Length:500 mm
- Seq:01
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Pk:15
Specifications
About this item
PE sleeves for storage in liquid nitrogen and to provide extra safety when freezing hazardous samples.
- Can be cut to the desired length
- Shrinks with gentle heating
External Ø×L: 14×500 mm