Orders LinkContactUsLinkComponent
Cryogenic tubes
Cryogenic tubes
Supplier:   Brand
Cryogenic tubes
Supplier:  Brand
Certificates
Cryogenic tubes
12 Options Available Below
Product Details & Documents
PP, graduated. Designed for storage of biological material, such as microorganisms, human and animal cells, more. in the gaseous phase of liquid nitrogen.
  • Large frosted marking area and coloured cap inserts for easy sample identification
  • Temperature stability to −196 °C and autoclavable at 121 °C (2 bar), acc. DIN EN 285
  • γ-ray sterile (SAL 10⁻⁶)
  • Cryogenic tubes are sterile, RNase-, DNase-, DNA- and endotoxin-free
  • Tubes without ring stands can be centrifuged at up to an RCF of 14000×g
Freestanding
* with round bottom
Product Family Options
With external thread
Product Information
  • Capacity
    Ø×H
    Cap included
    Availability
    Price
  • 1,20 ml
    12,5×41 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 427.00
    SFr. 427.00
    1 * 1.000 ST
       SFr. 427.00
  • 2,00 ml
    12,5×47 * mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 427.00
    SFr. 427.00
    1 * 1.000 ST
       SFr. 427.00
  • 2,00 ml
    12,5×49 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 598.00
    SFr. 598.00
    1 * 1.000 ST
       SFr. 598.00
  • 3,00 ml
    12,5×70 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 457.00
    SFr. 457.00
    1 * 1.000 ST
       SFr. 457.00
  • 4,00 ml
    12,5×76 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 495.00
    SFr. 495.00
    1 * 1.000 ST
       SFr. 495.00
  • 5,00 ml
    12,5×90 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 522.00
    SFr. 522.00
    1 * 1.000 ST
       SFr. 522.00
With internal thread
Product Information
  • Capacity
    Ø×H
    Cap included
    Availability
    Price
  • 1,20 ml
    12,5×41 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 427.00
    SFr. 427.00
    1 * 1.000 ST
       SFr. 427.00
  • 2,00 ml
    12,5×49 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 427.00
    SFr. 427.00
    1 * 1.000 ST
       SFr. 427.00
  • 2,00 ml
    12,5×48 * mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 427.00
    SFr. 427.00
    1 * 1.000 ST
       SFr. 427.00
  • 4,00 ml
    12,5×70 * mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 568.00
    SFr. 568.00
    1 * 1.000 ST
       SFr. 568.00
  • 4,00 ml
    12,5×71 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 647.00
    SFr. 647.00
    1 * 1.000 ST
       SFr. 647.00
  • 5,00 ml
    12,5×90 * mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 649.00
    SFr. 649.00
    1 * 1.000 ST
       SFr. 649.00
Cap inserts
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Blue
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 22.00
    SFr. 22.00
    1 * 500 ST
       SFr. 22.00
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Red
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 22.00
    SFr. 22.00
    1 * 500 ST
       SFr. 22.00
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 22.00
    SFr. 22.00
    1 * 500 ST
       SFr. 22.00
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Green
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 22.00
    SFr. 22.00
    1 * 500 ST
       SFr. 22.00
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Yellow
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 22.00
    SFr. 22.00
    1 * 500 ST
       SFr. 22.00