WHEATON® NextGen™ V Vial® Crimp/Screw Neck Vials
Fournisseur: DWK Life Sciences
Certificats
À propos de cet article
Clear vials conform to USP Type I and ASTM E 438 Type I, Class A requirements. Amber vials made from borosilicate glass that conforms to USP Type I requirements for light transmission to protect light-sensitive products. These vials are ideal for small-scale reactions, centrifugation, storage, packaging and shipping.
- Conical interior provides downward drainage for maximum sample retrieval
- Low particulate packaging protects against contamination during transportation and storage
Informations de commande: Available in screw thread or crimp / serum finish.
Frequently Bought Together