ContactUsLinkComponent
Tubing selector
Tubing selector
Catalog # 229-0111
Supplier:  KARTELL
undefined
Tubing selector
Catalog # 229-0111
Supplier:  KARTELL
Specifications
  • Description:
    Tubing selector
  • L×W:
    165×90 mm
  • Pk:
    1
Specifications
About this item
ABS, green.
  • Sturdy, handy shape
  • Engraved numbers for easy reading
For rapid determination of the internal and external diameters of tubing, glass tubes, glass rods, test tubes etc. For internal diameters from 1 to 25 mm and external diameters from 1,5 to 21 mm.