Orders LinkContactUsLinkComponent
Clamp gaskets, HelixMark®
Clamp gaskets, HelixMark®
Clamp gaskets, HelixMark®
Certificates
Clamp gaskets, HelixMark®
Clamp gaskets, HelixMark®
9 Options Available Below
Product Details & Documents
Silicone, platinum-cured.
  • Manufactured according to GMP in a production facility registered by the FDA for the manufacture of medical products
  • Can be sterilised by autoclaving, by ethylene oxide and via gamma radiation within the user's validation cycles and sterilisation levels
  • Meet USP Class VI requirements. Material certification and lot traceability are included in each box
These biomedical/pharmaceutical grade clamp gaskets promote crevice-free and protrusion-free processing. Contain no contaminants or leachables.
ISO 13485
Certifications: ISO 13485
Product Family Options
Product Information
  • Ø int.
    Availability
    Price
  • 12,70 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 443.00
    SFr. 443.00
    1 * 25 ST
       SFr. 443.00
  • 19,05 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 528.00
    SFr. 528.00
    1 * 25 ST
       SFr. 528.00
  • 25,40 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 689.00
    SFr. 689.00
    1 * 25 ST
       SFr. 689.00
  • 38,10 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 793.00
    SFr. 793.00
    1 * 25 ST
       SFr. 793.00
  • 50,80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 858.00
    SFr. 858.00
    1 * 25 ST
       SFr. 858.00
  • 63,50 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 937.00
    SFr. 937.00
    1 * 25 ST
       SFr. 937.00
  • 76,20 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 1’150.00
    SFr. 1’150.00
    1 * 25 ST
       SFr. 1’150.00
  • 101,60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 1’270.00
    SFr. 1’270.00
    1 * 25 ST
       SFr. 1’270.00
  • 152,40 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 3’050.00
    SFr. 3’050.00
    1 * 25 ST
       SFr. 3’050.00