Metal tubing with small flanges
Anbieter: VACUUBRAND
Zertifikate
Über diesen Artikel
Stainless steel.
- Meets strict gas discharge and cleanliness requirements
- Simple, easy assembly and disassembly
- Secure, vacuum tight connection in conjunction with optional centering and clamping rings
Highly flexible corrugated metal tubes with small flanges welded on both ends.
Technische Daten
- Catalog No:
- 185-2521
- 185-2511
- 185-2543
- 185-2532
- 185-2533
- 185-2522
- 185-2523
- 185-2544
- 185-2512
- 185-2513
- 185-2534
- 185-2541
- 185-2542
- 185-2524
- 185-2531
- 185-2514
- Repeated bending:
- 120 mm
- 90 mm
- 200 mm
- 155 mm
- 155 mm
- 120 mm
- 120 mm
- 200 mm
- 90 mm
- 90 mm
- 155 mm
- 200 mm
- 200 mm
- 120 mm
- 155 mm
- 90 mm
- Single bending:
- 29 mm
- 19 mm
- 65 mm
- 43 mm
- 43 mm
- 29 mm
- 29 mm
- 65 mm
- 19 mm
- 19 mm
- 43 mm
- 65 mm
- 65 mm
- 29 mm
- 43 mm
- 19 mm
- Flange size:
- KF DN 16
- KF DN 10
- KF DN 40
- KF DN 25
- KF DN 25
- KF DN 16
- KF DN 16
- KF DN 40
- KF DN 10
- KF DN 10
- KF DN 25
- KF DN 40
- KF DN 40
- KF DN 16
- KF DN 25
- KF DN 10
Technische Daten
Frequently Bought Together