Pumpenschläuche, Tygon® F-4040-A, konfektioniert mit Stoppern
Anbieter: Saint Gobain Life Sciences
Zertifikate
Über diesen Artikel
Tygon® -Schlauch, gelblich, durchscheinend
- Reißfest, quellen nicht auf, verspröden nicht
- Ozon- und UV-beständig
- Kompatibel mit den meisten erdölbasierenden Produkten
Petroleum and lubricant resistant tubing, that is ideal for the transfer of fuel, kerosene, heating oil, cutting fluids and glycol-based coolants. Does not swell up and harden even when hydrocarbon based liquids are used. This minimises the risk of defects such as cracking or leakage. Due to the low extraction level, tubing also protects the fluids from contamination. Length 381 mm. Distance between stops: 140 mm (2 stops) and 170 mm (3 stops).
Temperature resistance: −35 to +74 °C
Härte (Shore A): 57
Dichte: 1,26
Frequently Bought Together