Order Entry
Switzerland
ContactUsLinkComponent
Sterilin™ Test Tubes
Katalognr. SLINRT15
Anbieter:  STERILIN
undefined
Sterilin™ Test Tubes
Katalognr. SLINRT15
Anbieter:  STERILIN

Technische Daten

  • Ext.Ø:
    6 mm
  • Sterility:
    Non sterile
  • Type:
    Base tube
  • Capacity:
    0,6 ml
  • Description:
    Round base plastic tubes
  • Length:
    40 mm
  • Material:
    Polystyrene
  • Seq:
    01
  • Pk:
    10.000

Technische Daten

Über diesen Artikel

Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic round base tubes are manufactured from virgin polystyrene and supplied non sterile. Not available in all geographic regions.

  • Convenient alternative to glass tubes
  • Suitable for samples ranging from 0,6 to 11 ml
Delivery: Supplied clean and dust free.