Orders LinkContactUsLinkComponent
Test tubes without rim, medium wall | Avantor
Test tubes without rim, medium wall
Supplier:   MURRAY S
Test tubes without rim, medium wall
Supplier:  MURRAY S
Certificates
Test tubes without rim, medium wall
Test tubes without rim, medium wall
14 Options Available Below
Product Details & Documents
Soda-lime glass, rimless.
  • Good chemical and physical properties
  • Medium-walled
Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.
Product Family Options
Product Information
  • Length
    Ext.Ø
    Availability
    Price
  • 50 mm
    6,5 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - SFr. 102.00
    SFr. 102.00
    1 * 250 ST
       SFr. 102.00
  • 50 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - SFr. 102.00
    SFr. 102.00
    1 * 250 ST
       SFr. 102.00
  • 50 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 44.90
    SFr. 44.90
    1 * 100 ST
       SFr. 44.90
  • 75 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 49.40
    SFr. 49.40
    1 * 100 ST
       SFr. 49.40
  • 75 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 46.20
    SFr. 46.20
    1 * 100 ST
       SFr. 46.20
  • 75 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 61.90
    SFr. 61.90
    1 * 100 ST
       SFr. 61.90
  • 90 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 54.30
    SFr. 54.30
    1 * 100 ST
       SFr. 54.30
  • 100 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 62.70
    SFr. 62.70
    1 * 100 ST
       SFr. 62.70
  • 100 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 64.80
    SFr. 64.80
    1 * 100 ST
       SFr. 64.80
  • 125 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 79.60
    SFr. 79.60
    1 * 100 ST
       SFr. 79.60
  • 125 mm
    18 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 133.00
    SFr. 133.00
    1 * 100 ST
       SFr. 133.00
  • 150 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 89.10
    SFr. 89.10
    1 * 100 ST
       SFr. 89.10
  • 150 mm
    18 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 106.00
    SFr. 106.00
    1 * 100 ST
       SFr. 106.00
  • 150 mm
    24 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 98.20
    SFr. 98.20
    1 * 50 ST
       SFr. 98.20
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..