Test tubes without rim, medium wall
Supplier: MURRAY S
Jump to:
About this item
Borosilicate glass, grade 3.3, SAMCO '33'. Medium walled.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
Certifications: ISO 4142:2002
9 Options Available Below
Product Details & Documents
Borosilicate glass, grade 3.3, SAMCO '33'. Medium walled.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
Certifications: ISO 4142:2002
Product Family Options
Product Information
- LengthExt.ØAvailabilityPrice
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..