Orders LinkContactUsLinkComponent
Test tubes without rim
Test tubes without rim
Test tubes without rim
Certificates
Test tubes without rim
Test tubes without rim
15 Options Available Below
Product Details & Documents
AR-Glas® (soda-lime glass).
  • Heavy wall (212-1700 suitable as Durham-Tube)
  • Good chemical and physical properties
  • Can also be used as disposable test tubes
Documents
Product Family Options
Product Information
  • Length
    Ext.Ø
    Wall thickness
    Availability
    Price
  • 35 mm
    6 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 13.00
    SFr. 13.00
    1 * 100 ST
       SFr. 13.00
  • 40 mm
    8 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 74.70
    SFr. 74.70
    1 * 500 ST
       SFr. 74.70
  • 50 mm
    6,5 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 86.50
    SFr. 86.50
    1 * 500 ST
       SFr. 86.50
  • 70 mm
    10,5 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 14.90
    SFr. 14.90
    1 * 100 ST
       SFr. 14.90
  • 75 mm
    12 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - SFr. 22.70
    SFr. 22.70
    1 * 200 ST
       SFr. 22.70
  • 80 mm
    10,5 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - SFr. 54.60
    SFr. 54.60
    1 * 200 ST
       SFr. 54.60
  • 85 mm
    15 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 27.00
    SFr. 27.00
    1 * 100 ST
       SFr. 27.00
  • 100 mm
    10 mm
    0,9 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 24.50
    SFr. 24.50
    1 * 100 ST
       SFr. 24.50
  • 100 mm
    11 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 15.20
    SFr. 15.20
    1 * 100 ST
       SFr. 15.20
  • 100 mm
    13 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 23.90
    SFr. 23.90
    1 * 100 ST
       SFr. 23.90
  • 100 mm
    16 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 19.70
    SFr. 19.70
    1 * 100 ST
       SFr. 19.70
  • 130 mm
    14 mm
    0,9 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 28.20
    SFr. 28.20
    1 * 100 ST
       SFr. 28.20
  • 135 mm
    14,5 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 31.50
    SFr. 31.50
    1 * 100 ST
       SFr. 31.50
  • 160 mm
    16 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST
    Specifications:
    New Product
    New Product
  • 180 mm
    18 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 43.70
    SFr. 43.70
    1 * 100 ST
       SFr. 43.70