Centrifuge tubes
Anbieter: VITLAB
Zertifikate
Über diesen Artikel
PFA.
- Translucent
- Withstand forces up to 10000×g
- Also for autosampler racks
Ordering information: 525-2251 without screw cap, 525-2250 with calibrated ring-mark at 10 ml, thread GL25 and screw cap.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together