Centrifuge tubes, Oak Ridge, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Round bottom and rimless, autoclavable.
- Withstands forces up to 50000×g
- Suitable for use in refrigerated or non refrigerated centrifuges
- At speeds greater than 10000×g, sealing cap assemblies are recommended
FEP
Translucent. Resists any acid, alkali or solvent. Excellent for chloroform and phenol extractions.
PSF
Transparent. Acid and alkali resistant.
PC
Transparent.
PPCO
Translucent. Greater chemical resistance than PC.
Caution: Note: (Actual brim capacity of 50 ml Centrifuge tubes Oak Ridge is 42 ml).
Specifications
- Catalog No:
- 525-2224
- 525-2223
- 525-2213
- 525-2212
- 525-2211
- 525-2233
- 525-0046
- 525-0045
- 525-0244
- 525-2244
- 525-0044
- 525-2243
- 525-0049
- 525-2236
- 525-0042
- 525-0038
- 525-0043
- 525-2241
- 525-2235
- 525-2234
- 525-0037
- 525-0041
- 525-0040
- 525-2231
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement