ContactUsLinkComponent
Trolley, folding
Trolley, folding
Catalog # 139-9874
undefined
Trolley, folding
Catalog # 139-9874
Specifications
  • W×D×H:
    280×260×100 mm
  • Pk:
    1
Specifications
About this item
All purpose laboratory cart for all types of load, such as boxes, sacks, or drums. Easy to manoeuvre, even in confined spaces. Has a folding base plate to minimise storage space.
  • Solid steel tube frame
  • Solid rubber wheels with 125 mm