FluoroTrans® PVDF Transfer Membranes
About this item
Naturally hydrophobic PVDF, ideal for a wide variety of protein-analysis applications. The family of FluroTrans® media are white, microporous solid phase supports that bind proteins tenaciously via hydrophobic interactions. FluoroTrans® membrane is optimised for N-terminal protein sequencing. FluoroTrans® W membranes are optimised for Western transfer applications. They allow for sensitive protein detection with low background and very low protein burn-through. Immobilised proteins can be visually detected with Coomassie® Blue, Amido black, Ponceau S, and colloidal gold.
- High protein binding capacity - typically absorbs 50% more protein than nylon or nitrocellulose
- High tensile strength, so won't tear, crack or curl during handling, allowing easy removal of target bands for protein sequencing applications
- High sensitivity for small peptides
Pore size: 0,2 µm
Typical thickness: 127 µm
8 Options Available Below
Product Details & Documents
Naturally hydrophobic PVDF, ideal for a wide variety of protein-analysis applications. The family of FluroTrans® media are white, microporous solid phase supports that bind proteins tenaciously via hydrophobic interactions. FluoroTrans® membrane is optimised for N-terminal protein sequencing. FluoroTrans® W membranes are optimised for Western transfer applications. They allow for sensitive protein detection with low background and very low protein burn-through. Immobilised proteins can be visually detected with Coomassie® Blue, Amido black, Ponceau S, and colloidal gold.
- High protein binding capacity - typically absorbs 50% more protein than nylon or nitrocellulose
- High tensile strength, so won't tear, crack or curl during handling, allowing easy removal of target bands for protein sequencing applications
- High sensitivity for small peptides
Pore size: 0,2 µm
Typical thickness: 127 µm
Documents
VWR® Products for Electrophoresis
Product Literature
Product Family Options
FluoroTrans® membranes
Product Information
- DimensionsAvailabilityPrice
- 516-0220PVDF transfer membranes, sheets, pore size 0,2 µmPVDF transfer membranes, sheets, pore size 0,2 µm85×90 mmLog in for availability1 * 20 STSFr. 191.00
- 516-0222PVDF transfer membranes, sheets, pore size 0,2 µmPVDF transfer membranes, sheets, pore size 0,2 µm130×140 mmLog in for availability1 * 10 STSFr. 556.00
- 516-9757PVDF transfer membranes, roll, pore size 0,2 µmPVDF transfer membranes, roll, pore size 0,2 µm260×3000 mmLog in for availability1 * 1 ROUSFr. 509.00
- 516-0224PVDF transfer membranes, roll, pore size 0,2 µmPVDF transfer membranes, roll, pore size 0,2 µm300×3300 mmLog in for availability1 * 1 ROUSFr. 484.00
FluoroTrans® W membranes
Product Information
- DimensionsAvailabilityPrice
- 516-0225PVDF transfer membranes, sheets, pore size 0,2 µmPVDF transfer membranes, sheets, pore size 0,2 µm100×150 mmLog in for availability1 * 10 STSFr. 276.00
- 516-0248PVDF transfer membranes, sheets, pore size 0,2 µmPVDF transfer membranes, sheets, pore size 0,2 µm70×90 mmLog in for availability1 * 10 STSFr. 198.00
- 516-0226PVDF transfer membranes, sheets, pore size 0,2 µmPVDF transfer membranes, sheets, pore size 0,2 µm200×200 mmLog in for availability1 * 10 STSFr. 433.00
- 516-0249PVDF transfer membranes, roll, pore size 0,2 µmPVDF transfer membranes, roll, pore size 0,2 µm300×3300 mmLog in for availability1 * 1 ROUSFr. 610.00