Order Entry
Switzerland
ContactUsLinkComponent
Safety cutter, SECUNORM MULTISAFE
Safety cutter, SECUNORM MULTISAFE
  233-0196
undefined
Safety cutter, SECUNORM MULTISAFE
  233-0196

 

  • Total length:
    142 mm
  • Blade:
    53×19×0,63 mm
  • Pk:
    1

 

 

Trapezoid blade, handle made of solid die-cast zinc.

  • Full metal housing: very robust and abrasion resistant
  • Interchangeable blade (tool required)
  • With spring-loaded blade retraction and blade control slider on top
  • Trapezoid deep-edged installed blade
  • Blade with two cutting edges doubles the efficiency of the cutter

Heavy-duty cutter for use by left and right-handed users. The spring-loaded blade retract system is designed to respond to any uncontrolled move if and when the thumb-push slider is not in forward position. Ideal for cutting tape, plastic, cardboard, textile, rubber, PVC and leather.

TÜV approved

 : TÜV approved