Orders LinkContactUsLinkComponent
General purpose thermometers for flasks
General purpose thermometers for flasks
Supplier:   Amarell
General purpose thermometers for flasks
Supplier:  Amarell
Certificates
General purpose thermometers for flasks
General purpose thermometers for flasks
2 Options Available Below
Product Details & Documents
These strengthened in-glass thermometers provides reinforced tips for stirring applications.
  • Fitted anti-roll plastic cap with cord loop
  • White enamelled back
  • No breakable glass ring
  • Bulb integral with stem
  • 620-0121 and 620-0122 calibrated at four and five points respectively
Mercury.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Product Family Options
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • ME9010
    −10...+50 °C
    0,5 °C
    305 mm
    Mercury
    76 mm
    6,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • ME9008
    −10...+110 °C
    1,0 °C
    305 mm
    Mercury
    76 mm
    6,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST