ContactUsLinkComponent
Protective film, Bytac®
  293-4331
undefined
Protective film, Bytac®
  293-4331
  • Packed:
    Roll
  • Colour:
    White
  • L×W:
    4 600×635 mm
  • Pk:
    4,6 m
Self-adhesive laminate, consisting of a chemical-resistant FEP film surface and a vinyl substrate for protecting all common work surfaces. Can be easily peeled away without leaving any adhesive residue.
Temperature resistance: Vinyl: Up to a maximum of 93 °C ; Aluminium: Up to 182 °C (at a maximum of one hour).