Orders LinkContactUsLinkComponent
Stopcocks, one-way
Stopcocks, one-way
Supplier:   Lenz Laborglas
Stopcocks, one-way
Supplier:  Lenz Laborglas
Certificates
Stopcocks, one-way
Stopcocks, one-way
14 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
Certifications: DIN 12554.
Documents
Product Family Options
With solid glass key
Product Information
  • Hole Ø
    Spigot
    Availability
    Price
  • 1,5 mm
    12,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 38.40
    SFr. 38.40
    1 * 1 ST
       SFr. 38.40
  • 2,5 mm
    12,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 38.40
    SFr. 38.40
    1 * 1 ST
       SFr. 38.40
  • 2,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 40.10
    SFr. 40.10
    1 * 1 ST
       SFr. 40.10
  • 4,0 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 45.80
    SFr. 45.80
    1 * 1 ST
       SFr. 45.80
  • 4,0 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 58.20
    SFr. 58.20
    1 * 1 ST
       SFr. 58.20
  • 6,0** mm
    21,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 72.70
    SFr. 72.70
    1 * 1 ST
       SFr. 72.70
  • 8,0** mm
    24,0 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 122.00
    SFr. 122.00
    1 * 1 ST
       SFr. 122.00
  • 10** mm
    29,2 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 124.00
    SFr. 124.00
    1 * 1 ST
       SFr. 124.00
With PTFE key
Product Information
  • Hole Ø
    Spigot
    Availability
    Price
  • 1,5 mm
    12,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 45.40
    SFr. 45.40
    1 * 1 ST
       SFr. 45.40
  • 2,5 mm
    12,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 47.70
    SFr. 47.70
    1 * 1 ST
       SFr. 47.70
  • 2,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 50.60
    SFr. 50.60
    1 * 1 ST
       SFr. 50.60
  • 4,0 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 57.40
    SFr. 57.40
    1 * 1 ST
       SFr. 57.40
  • 4,0 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 73.20
    SFr. 73.20
    1 * 1 ST
       SFr. 73.20
  • 6,0 mm
    21,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 105.00
    SFr. 105.00
    1 * 1 ST
       SFr. 105.00