Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

PP, 6 mm, 244 mm
Catalog # 442-4160
Supplier:   KARTELL
Stirring rods
Catalog # 442-4160
Supplier:  KARTELL
Stirring rods
Stirring rods

PP, 6 mm, 244 mm

Price
SFr. 15.90
Log in for availability
1 * 10 ST
Product Details & Documents

White, one-piece rod with flattened ends which form two small spatulas.

  • Dimensions of spatulas (W×H) 15×12 and 12×9 mm

Dimensions of spatulas (W×H) 15×12 and 12×9 mm.

Specifications
  • Ø:
    6 mm
  • Length:
    244 mm
  • Material:
    PP
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.