Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stirring rods, Nalgene®

Stirring rods, Nalgene®

Stirring rods, PP, PP, 6,4 mm, 248 mm
Catalog # 231-0074
   Low Manufacturing Impact
Stirring rods, Nalgene®
Catalog # 231-0074
   Low Manufacturing Impact
Stirring rods, Nalgene®
Stirring rods, Nalgene®

Stirring rods, PP, PP, 6,4 mm, 248 mm

Price
SFr. 114.00
Log in for availability
1 * 12 ST
Product Details & Documents

Stirring rod with flattened ends forming flat paddle 16×16 mm at one end and triangular paddle at the other end.

  • Good chemical resistance
  • Autoclavable
Specifications
  • Ø:
    6,4 mm
  • Description:
    Stirring rods, PP
  • Length:
    248 mm
  • Material:
    PP
  • Cat. No.:
    231-0074
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.