ContactUsLinkComponent
Adapters, steam distillation head, Quickfit®
Adapters, steam distillation head, Quickfit®
Codice catalogo BIBBSH7/23/SC
Nome del fornitore:  SciLabware
Adapters, steam distillation head, Quickfit®
Codice catalogo BIBBSH7/23/SC
Nome del fornitore:  SciLabware
Specifiche
  • Cone to condenser:
    19/26 NS
  • Cone to flask:
    24/29 NS
  • Seq:
    01
  • Pk:
    5
Specifiche
Informazioni su questo articolo
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
A splash-head with integral stem for the introduction of steam into flasks. Stem is connected to steam supply via detachable screwthread safety connector.