ContactUsLinkComponent
Distilling bridges, Quickfit®
Distilling bridges, Quickfit®
Catalog # 201-8692
Supplier:  SciLabware
undefined
Distilling bridges, Quickfit®
Catalog # 201-8692
Supplier:  SciLabware
Specifications
  • Cone size:
    24/29 NS
  • Pk:
    1
Specifications
About this item
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Used in destillation assemblies in which the condenser is assembled in the vertical mode.