Spore discs, Sportrol™
: CROSSTEX
Biological sterilisation discs are used to perform sterilisation cycle validation in small areas of a device, where a standard-sized spore strip will not fit, such as within a syringe barrel, inside tubing, or under a cap. Spore discs are created by inoculating spore suspensions of certified population and resistance onto paper discs.
- Spore population: 106 spores per disc
- Disc diameter: 6,35 mm
- 24-month shelf life when stored under labelled conditions
: Meet USP and ISO 11138-1 standards.
: Packaged in bulk, in shelf packs of 100. Supplied with instructions for use, certified population, D-value, Z-value, survival/kill data, and expiry date.
Frequently Bought Together