VWR® PUREImage, Cleanroom Papers
Cleanroom paper is designed for use in cleanroom environments, where the prevention of chemical and particle contamination is critical.
- High tensile strength, and excellent heat resistance, for enhanced copier and printing efficiency
- PE-binding technology results in very low levels of particles, non volatile residue and ion extractables
- Unique precision-cutting technology guarantees consistent sizing and cleanliness
- Paper is web-cleaned on both sides prior to sheeting
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double-bagged in an ISO Class 5 (FED-STD-209E Class 100) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10) to ISO Class 6 (FED-STD-209E Class 1000) cleanrooms.
Weight: 80 g/m², except 115-0156 and 115-0148 with 105 g/m²
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double bagged in an ISO Class 5 (FED-STD-209E Class 100/M3,5) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10/M2,5) to ISO Class 6 (FED-STD-209E Class 1000/M4,5) cleanrooms.
: Paper is packed 250 sheets per pack, 10 packs per case.
- Catalog No:
- 115-1961
- 115-1959
- 115-0149
- 115-1957
- 115-1958
- 115-1960
- 115-0150
- 115-0156
- 115-1951
- 115-1953
- 115-0164
- 115-1952
- 115-0163
- 115-1956
- 115-1955
- 115-0166
- 115-1954
- 115-0165
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
Frequently Bought Together