Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Micro powder spoon

Micro powder spoon

150 mm, 5 mm
Catalog # 231-0099
Supplier:   RSG
Micro powder spoon
Catalog # 231-0099
Supplier:  RSG
Micro powder spoon
Micro powder spoon

150 mm, 5 mm

Price
23.10 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. With hexagonal handle for easier handling.

  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Length:
    150 mm
  • Spoon size:
    5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.