Order Entry
Switzerland
ContactUsLinkComponent
Vibration spatula
Vibration spatula
Catalog # 231-1289
Supplier:  BOCHEM
undefined
Vibration spatula
Catalog # 231-1289
Supplier:  BOCHEM

Specifications

  • Length:
    190 mm
  • Blade width:
    12 mm
  • Pk:
    1
  • Blade length:
    50 mm

Specifications

About this item

Stainless steel, plastic shaft. A toothed wheel is turned by a finger making the spatula blade vibrate. This action allows for accurate adding of material.