Order Entry
Switzerland
ContactUsLinkComponent
Trough spatula
Trough spatula
  231-0172
undefined
Trough spatula
  231-0172

 

  • Length:
    150 mm
  • Pk:
    10

 

 

Stainless steel (304), non magnetic.

  • Double-ended: One flat end, one trough bladed end
  • Superior corrosion resistance and durability

Trough bladed end: 8 mm wide × 50 mm long × 1,5 mm deep