Pharmacist's spatulas
: RSG
Chromium steel, hardened, extremely flexible blade.
- Excellent thermal resistance
- Good chemical resistance
- Quick and easy maintenance
- Available with wooden handle or with dishwasher-safe plastic handle
Frequently Bought Together