Double spatulas, micro
Supplier: RSG
Jump to:
About this item
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- Both ends have an angled spoon
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
7 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- Both ends have an angled spoon
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
Product Family Options
Product Information
- LengthBlade widthAvailabilityPrice