Orders LinkContactUsLinkComponent
Spoon spatulas, micro
Spoon spatulas, micro
Supplier:   RSG
Spoon spatulas, micro
Supplier:  RSG
Certificates
Spoon spatulas, micro
Spoon spatulas, micro
9 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • With a micro spoon at one end and a micro spatula at the other
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
Product Family Options
Product Information
  • Length
    Blade width
    Availability
    Price
  • 210 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 47.60
    SFr. 47.60
    1 * 5 ST
       SFr. 47.60
  • 210 mm
    7 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 47.80
    SFr. 47.80
    1 * 5 ST
       SFr. 47.80
  • 185 mm
    7 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 47.60
    SFr. 47.60
    1 * 5 ST
       SFr. 47.60
  • 150 mm
    7 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 46.40
    SFr. 46.40
    1 * 5 ST
       SFr. 46.40
  • 400 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 61.60
    SFr. 61.60
    1 * 5 ST
       SFr. 61.60
  • 185 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 47.00
    SFr. 47.00
    1 * 5 ST
       SFr. 47.00
  • 100 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 45.50
    SFr. 45.50
    1 * 5 ST
       SFr. 45.50
  • 150 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 46.00
    SFr. 46.00
    1 * 5 ST
       SFr. 46.00
  • 300 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 54.20
    SFr. 54.20
    1 * 5 ST
       SFr. 54.20