Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
Supplier:   BOCHEM
Double spatulas, micro
Supplier:  BOCHEM
Certificates
Double spatulas, micro
Double spatulas, micro
26 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • With round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Product Family Options
Product Information
  • Length
    Blade length
    Blade width
    Availability
    Price
  • 100 mm
    30 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2.70
    SFr. 2.70
    1 * 1 ST
       SFr. 2.70
  • 100 mm
    35 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 13.10
    SFr. 13.10
    1 * 5 ST
       SFr. 13.10
  • 100 mm
    35 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2.70
    SFr. 2.70
    1 * 1 ST
       SFr. 2.70
  • 100 mm
    35 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2.70
    SFr. 2.70
    1 * 1 ST
       SFr. 2.70
  • 100 mm
    35 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2.70
    SFr. 2.70
    1 * 1 ST
       SFr. 2.70
  • 130 mm
    40 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.00
    SFr. 3.00
    1 * 1 ST
       SFr. 3.00
  • 130 mm
    40 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 15.10
    SFr. 15.10
    1 * 5 ST
       SFr. 15.10
  • 130 mm
    40 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.00
    SFr. 3.00
    1 * 1 ST
       SFr. 3.00
  • 130 mm
    40 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.00
    SFr. 3.00
    1 * 1 ST
       SFr. 3.00
  • 130 mm
    40 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.00
    SFr. 3.00
    1 * 1 ST
       SFr. 3.00
  • 150 mm
    40 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.20
    SFr. 3.20
    1 * 1 ST
       SFr. 3.20
  • 150 mm
    40 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.20
    SFr. 3.20
    1 * 1 ST
       SFr. 3.20
  • 150 mm
    40 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.20
    SFr. 3.20
    1 * 1 ST
       SFr. 3.20
  • 150 mm
    40 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 16.00
    SFr. 16.00
    1 * 5 ST
       SFr. 16.00
  • 150 mm
    40 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.20
    SFr. 3.20
    1 * 1 ST
       SFr. 3.20
  • 150 mm
    40 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.20
    SFr. 3.20
    1 * 1 ST
       SFr. 3.20
  • 150 mm
    40 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.20
    SFr. 3.20
    1 * 1 ST
       SFr. 3.20
  • 185 mm
    50 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.30
    SFr. 3.30
    1 * 1 ST
       SFr. 3.30
  • 185 mm
    50 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 17.00
    SFr. 17.00
    1 * 5 ST
       SFr. 17.00
  • 185 mm
    50 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.40
    SFr. 3.40
    1 * 1 ST
       SFr. 3.40
  • 185 mm
    50 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.30
    SFr. 3.30
    1 * 1 ST
       SFr. 3.30
  • 185 mm
    50 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.40
    SFr. 3.40
    1 * 1 ST
       SFr. 3.40
  • 210 mm
    55 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 17.50
    SFr. 17.50
    1 * 5 ST
       SFr. 17.50
  • 210 mm
    55 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.60
    SFr. 3.60
    1 * 1 ST
       SFr. 3.60
  • 210 mm
    55 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.60
    SFr. 3.60
    1 * 1 ST
       SFr. 3.60
  • 210 mm
    55 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.60
    SFr. 3.60
    1 * 1 ST
       SFr. 3.60