Sealing film and foil for microplates, Whatman™
512-1087
- Sterile:-
- Description:Cold sealing film
- Material:Polyester
- Thickness:50 µm
- Pk:100
Seals are used to control humidity and reduce evaporation of samples. They prevent spills and contamination.
- Cold seals in clear polyester or PP are self-sticking with inert adhesive
- Heat seals are suitable for PP microplates only and are applied using heat and pressure