Sealing film, reflective, for PCR plates, Axygen®
: Corning
This reflective aluminium film is ideal for sealing plates containing light sensitive samples. Seals during the thermo cycling process, when used in conjunction with a compression mat. Also provides an excellent seal when used with other polystyrene and polypropylene microplates.
- Provides exceptional seal under the widest temperature conditions, −80 to +104 °C
- Pierceable to allow for direct sample recovery without peeling
- Proprietary adhesive is inert and chemically resistant to DMSO
Frequently Bought Together