Orders LinkContactUsLinkComponent
Weighing scoops
Weighing scoops
Supplier:   Lenz Laborglas
Weighing scoops
Supplier:  Lenz Laborglas
Certificates
Weighing scoops
3 Options Available Below
Product Details & Documents
Borosilicate glass.
  • Ideal for weighing small samples
  • The tubular stem of the scoop also works as a weighing funnel and facilitates emptying of the scoop
Documents
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 6 ml
    95 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 37.30
    SFr. 37.30
    1 * 1 ST
       SFr. 37.30
  • 10 ml
    116 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 39.30
    SFr. 39.30
    1 * 1 ST
       SFr. 39.30
  • 3 ml
    82 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 33.50
    SFr. 33.50
    1 * 1 ST
       SFr. 33.50