Dippers
300 ml, 381 (Handle) mm
Catalog # NICK5072
Supplier: NICKEL ELECTRO
Jump to:
Product Details & Documents
Stainless steel. With extra-long handles. For milk sample collection and a wide range of other laboratory and industrial applications.
- Excellent chemical resistance
- Excellent thermal resistance
- Smooth surfaces for simple, easy cleaning
Specifications
- Capacity:300 ml
- Length:381 (Handle) mm