Orders LinkContactUsLinkComponent
Scoops
Supplier:   SCHROEDER WILESCO
Scoops
Supplier:  SCHROEDER WILESCO
Certificates
Scoops
10 Options Available Below
Product Details & Documents
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 50 ml
    135 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2.30
    SFr. 2.30
    1 * 1 ST
       SFr. 2.30
  • 90 ml
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.50
    SFr. 4.50
    1 * 1 ST
       SFr. 4.50
  • 160 ml
    210 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 5.10
    SFr. 5.10
    1 * 1 ST
       SFr. 5.10
  • 240 ml
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 7.10
    SFr. 7.10
    1 * 1 ST
       SFr. 7.10
  • 480 ml
    310 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.30
    SFr. 12.30
    1 * 1 ST
       SFr. 12.30
  • 900 ml
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 15.80
    SFr. 15.80
    1 * 1 ST
       SFr. 15.80
  • 1600 ml
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 22.40
    SFr. 22.40
    1 * 1 ST
       SFr. 22.40
  • 2500 ml
    430 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 31.20
    SFr. 31.20
    1 * 1 ST
       SFr. 31.20
  • 33 ml
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1.90
    SFr. 1.90
    1 * 1 ST
       SFr. 1.90
  • 85 ml
    155 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2.20
    SFr. 2.20
    1 * 1 ST
       SFr. 2.20