VWR®, Curved Base Stand
Catalog # 241-0104
Supplier: VWR Collection
Specifications
- Description:Curved base stand
- Environmentally Preferable:
- Pk:1
Specifications
About this item
PU coated cast iron stand, high grade steel rod 500×12 mm.
- Special shape of the stand ensures it is particularly solid and steady
The rod can be removed, has M8 thread, and is fixed from below with a washer and a nut.