Orders LinkContactUsLinkComponent
Tripod stands
Tripod stands
Supplier:   USBECK Laborgeräte
Tripod stands
Certificates
Tripod stands
4 Options Available Below
Product Details & Documents
Steel, powder coated.
  • With M10 threaded hole
  • No support rod
  • Non-slip, rubber feet ensure firm footing
Product Family Options
Product Information
  • Weight
    Arm length
    Availability
    Price
  • 241-0245
    Retort stand base, tripod
    Retort stand base, tripod
    1,0 kg
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 37.40
    SFr. 37.40
    1 * 1 ST
       SFr. 37.40
  • 241-0246
    Retort stand base, tripod
    Retort stand base, tripod
    1,35 kg
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 47.50
    SFr. 47.50
    1 * 1 ST
       SFr. 47.50
  • 241-0341
    Retort stand base, tripod
    Retort stand base, tripod
    1,7 kg
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 56.10
    SFr. 56.10
    1 * 1 ST
       SFr. 56.10
  • 241-0377
    Retort stand base, tripod
    Retort stand base, tripod
    0,68 kg
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 32.40
    SFr. 32.40
    1 * 1 ST
       SFr. 32.40