Laboratory stands
PP stand bases with rods made from chrome plated steel. Ideal for use as stands for burettes, funnels or separating funnels. Not recommended for heavier weights.
- Rods are 250 mm long, 12 mm diameter and can be screwed together to reach the desired height
- Model A: Stand base 300×150×15 mm with centre hole
- Model B: Stand base 300×150×15 mm with off-centre hole
Ordering information: Each pack includes two rods measuring 250×12 mm. Extra rods can be ordered separately.
PP stand bases with rods made from chrome plated steel. Ideal for use as stands for burettes, funnels or separating funnels. Not recommended for heavier weights.
- Rods are 250 mm long, 12 mm diameter and can be screwed together to reach the desired height
- Model A: Stand base 300×150×15 mm with centre hole
- Model B: Stand base 300×150×15 mm with off-centre hole
Ordering information: Each pack includes two rods measuring 250×12 mm. Extra rods can be ordered separately.