Orders LinkContactUsLinkComponent
Retort stands
Retort stands
Supplier:   USBECK Laborgeräte
Retort stands
Certificates
Retort stands
2 Options Available Below
Product Details & Documents
Stainless steel. Can be used for a wide range of equipment, e.g. stirrers, hotplates etc.
  • Model A: Consists of a base, 10 mm thick, 315×210 mm, and a rod, 800 mm long, 16 mm diameter
  • Model B: Consists of a base, 10 mm thick, 315×210 mm, and a rod, 1000 mm long, 16 mm diameter
  • Robust, firm, corrosion resistant, non slip
  • With three fixed feet and one adjustable
  • Threaded off-centre hole
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • 241-5121
    Model A
    Model A
    315×210 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 200.00
    SFr. 200.00
    1 * 1 ST
       SFr. 200.00
  • 241-5122
    Model B
    Model B
    315×210 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 206.00
    SFr. 206.00
    1 * 1 ST
       SFr. 206.00