Order Entry
Switzerland
ContactUsLinkComponent
VWR® Stand with Holder for Separating Funnels
VWR® Stand with Holder for Separating Funnels
Catalog # 241-0046
Supplier:  VWR Collection
undefined
VWR® Stand with Holder for Separating Funnels
Catalog # 241-0046
Supplier:  VWR Collection

Specifications

  • Description:
    Stand with funnel holder for two funnels
  • L×W:
    200×450 mm
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

PU coated cast iron stand 200×450 mm, steel rod 500×12 mm, bosshead and holder for two funnels.

  • Height of holder can be adjusted
  • Holder measures 300×100 mm
  • Apertures for funnels measure 65 mm - centre distance 200 mm