Safety shields
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Jump to:
About this item
Protective shield made from acrylic glass to be used as splinter shield and splash guard.
- Strong and stable design
- Removable feet
2 Options Available Below
Product Details & Documents
Protective shield made from acrylic glass to be used as splinter shield and splash guard.
- Strong and stable design
- Removable feet
Product Family Options
Product Information