ContactUsLinkComponent
Racks for microcentrifuge tubes
Racks for microcentrifuge tubes
Catalog # 211-7301
Supplier:  HEATHROW
undefined
Racks for microcentrifuge tubes
Catalog # 211-7301
Supplier:  HEATHROW
Specifications
  • For tubes:
    0,2/0,5/1,5/2,0 ml
  • No. of Holes:
    18
  • Colour:
    Blue
  • W×D×H:
    Ø 82×60 mm
  • Pk:
    5
Specifications
About this item
Polyfoam.
  • For a variety of tubes
  • Floating rack
  • Beaker-sized rack comes with an optional handle