ContactUsLinkComponent
Rack for cryogenic vials, Nunc™
Rack for cryogenic vials, Nunc™
Catalog # 391-8531
   Low Manufacturing Impact
undefined
Rack for cryogenic vials, Nunc™
Catalog # 391-8531
   Low Manufacturing Impact
Specifications
  • For:
    0 - 4,5 ml (Ø 12,5 mm)
  • No. of Holes:
    40 (4×10)
  • Colour:
    Blue
  • W×D×H:
    202×102×18 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Pk:
    15
Specifications
About this item
Polyphenyleneoxide, autoclavable.
Allows one handed operation when used with starfoot vials.