Order Entry
Switzerland
ContactUsLinkComponent
HPTLC plates, silica gel 60, unmodified layers, aluminium sheets
HPTLC plates, silica gel 60, unmodified layers, aluminium sheets
Supplier:  Merck
Certificates
undefined
HPTLC plates, silica gel 60, unmodified layers, aluminium sheets
Supplier:  Merck
Certificates

Specifications

  • Catalog No:
  • 1.05556.0001
  • 1.05548.0001
  • 1.05547.0001
  • Adsorption detection limit:
  • 100 - 500 pg
  • 100 - 500 pg
  • 100 - 500 pg
  • Fluorescence detection limit:
  • 5 - 10 pg
  • 5 - 10 pg
  • 5 - 10 pg
  • Average particle size:
  • 5 - 6 µm
  • 5 - 6 µm
  • 5 - 6 µm
  • Particle size distribution:
  • 4 - 8 µm
  • 4 - 8 µm
  • 4 - 8 µm
  • Sample volume:
  • 0,1 - 0,5 µl
  • 0,1 - 0,5 µl
  • 0,1 - 0,5 µl
  • No. of samples per plate:
  • <36
  • <36
  • <36
  • Typical migration distance:
  • 30 - 60 mm
  • 30 - 60 mm
  • 30 - 60 mm
  • Typical plate height:
  • 12 µm
  • 12 µm
  • 12 µm
  • Typical separation time:
  • 3 - 20 min
  • 3 - 20 min
  • 3 - 20 min

Specifications

Related Information

Frequently Bought Together