Serological pipettes, MBL
Anbieter: SciLabware
Zertifikate
Über diesen Artikel
Soda-lime glass, graduated, type 4.
- Calibrated to deliver from any graduation line down to zero at the jet, with the last drop expelled by blowing
- Tooled suction ends accept cotton wool plugs
- Graduations in permanent amber stain
Certifications: ISO 835
Frequently Bought Together